CAS RN 54231-36-6
CAS RN 54231-36-6
IUPAC Name:
4-(3-chloropyridin-2-yl)morpholine
Synonyms:
4-(3-chloropyridin-2-yl)morpholine
Molecular Weight (g/mol):
198.65
Molecular Formula:
C9H11ClN2O
InChi Key:
KCKQIVLNKGDTGJ-UHFFFAOYSA-N
SMILES:
ClC1=C(N=CC=C1)N1CCOCC1
1
–
1
of
1
results