accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 5371-70-0

CH3OOClNOOH3C

CAS RN 5371-70-0

IUPAC Name: 2,6-dimethyl 4-chloropyridine-2,6-dicarboxylate
Synonyms: 2,6-dimethyl 4-chloropyridine-2,6-dicarboxylate
Molecular Weight (g/mol): 229.62
Molecular Formula: C9H8ClNO4
InChi Key: NFXKYKHKNUFOKB-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=CC(=N1)C(=O)OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.