CAS RN 53609-64-6
CAS RN 53609-64-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express N-Bis(2-hydroxypropyl)nitrosamine, MedChemExpress
MedChemExpress N-Bis(2-hydroxypropyl)nitrosamine is an agent with carcinogenic activity.
