accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 511-67-1

BrBrONHOHNO

CAS RN 511-67-1

IUPAC Name: 5,5-dibromo-1,3-diazinane-2,4,6-trione
Synonyms: 5,5-dibromo-1,3-diazinane-2,4,6-trione
Molecular Weight (g/mol): 285.88
Molecular Formula: C4H2Br2N2O3
InChi Key: AMATXUCYHHHHHB-UHFFFAOYSA-N
SMILES: BrC1(Br)C(=O)NC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.