accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 452342-67-5

ONHONNHNN

CAS RN 452342-67-5

IUPAC Name: N-(oxan-4-yl)-4-{4-[5-(pyridin-2-yl)-1H-pyrazol-4-yl]pyridin-2-yl}benzamide
Synonyms: N-(oxan-4-yl)-4-{4-[3-(pyridin-2-yl)-2H-pyrazol-4-yl]pyridin-2-yl}benzamide
Molecular Weight (g/mol): 425.49
Molecular Formula: C25H23N5O2
InChi Key: SAGZIBJAQGBRQA-UHFFFAOYSA-N
SMILES: O=C(NC1CCOCC1)C1=CC=C(C=C1)C1=NC=CC(=C1)C1=C(NN=C1)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.