accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 368870-06-8

NOClOHO

CAS RN 368870-06-8

IUPAC Name: 1-[2-(4-chlorophenyl)ethyl]-5-oxopyrrolidine-3-carboxylic acid
Synonyms: 1-[2-(4-chlorophenyl)ethyl]-5-oxopyrrolidine-3-carboxylic acid
Molecular Weight (g/mol): 267.71
Molecular Formula: C13H14ClNO3
InChi Key: IMVZABZPKCETQX-UHFFFAOYNA-N
SMILES: OC(=O)C1CN(CCC2=CC=C(Cl)C=C2)C(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.