accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 349085-82-1

CH3H3CNHSOOOCH3CH3

CAS RN 349085-82-1

IUPAC Name: 4-methoxy-N-(2,4,6-trimethylphenyl)benzene-1-sulfonamide
Synonyms: 4-methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide
Molecular Weight (g/mol): 305.39
Molecular Formula: C16H19NO3S
InChi Key: FQUAFMNPXPXOJE-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)S(=O)(=O)NC1=C(C)C=C(C)C=C1C
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (5)

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.