accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 2554-06-5

H3CSiOSiCH3OSiCH3OSiH3COH2CH2CCH2CH2

CAS RN 2554-06-5

IUPAC Name: 2,4,6,8-tetraethenyl-2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
Synonyms: 2,4,6,8-tetraethenyl-2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
Molecular Weight (g/mol): 344.66
Molecular Formula: C12H24O4Si4
InChi Key: VMAWODUEPLAHOE-UHFFFAOYSA-N
SMILES: C[Si]1(O[Si](C)(O[Si](C)(O[Si](C)(O1)C=C)C=C)C=C)C=C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.