accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 19845-69-3

PP

CAS RN 19845-69-3

IUPAC Name: [6-(diphenylphosphanyl)hexyl]diphenylphosphane
Synonyms: [6-(diphenylphosphanyl)hexyl]diphenylphosphane
Molecular Weight (g/mol): 454.53
Molecular Formula: C30H32P2
InChi Key: GPORFKPYXATYNX-UHFFFAOYSA-N
SMILES: C(CCCP(C1=CC=CC=C1)C1=CC=CC=C1)CCP(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.