accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 19368-18-4

CH3H3CHNOOHO

CAS RN 19368-18-4

IUPAC Name: 2-[(2,6-dimethylphenyl)carbamoyl]benzoic acid
Synonyms: 2-[(2,6-dimethylphenyl)carbamoyl]benzoic acid
Molecular Weight (g/mol): 269.3
Molecular Formula: C16H15NO3
InChi Key: LLECMGGNFBKPRH-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1NC(=O)C1=CC=CC=C1C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.