CAS RN 1927857-56-4
CAS RN 1927857-56-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express PQR620, MedChemExpress
MedChemExpress PQR620 is an orally bioavailable and selective brain penetrant inhibitor of mTORC1/2.
