CAS RN 174022-42-5
CAS RN 174022-42-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Bevirimat, MedChemExpress
MedChemExpress Bevirimat (PA-457; MPC-4326; YK FH312) is an anti-HIV drug derived from a betulinic acid-like compound; is believed to inhibit HIV by a novel mechanism, so-called maturation inhibition.
