accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 17119-15-2

OHOHOHO

CAS RN 17119-15-2

IUPAC Name: 2-hydroxy-2-(3-hydroxyphenyl)acetic acid
Synonyms: MHMA M-hydroxymandelic acid M-hydroxymandelate
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: OLSDAJRAVOVKLG-UHFFFAOYNA-N
SMILES: OC(C(O)=O)C1=CC=CC(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.