CAS RN 17008-69-4
CAS RN 17008-69-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress Pseudobufarenogin
MedChemExpress Pseudobufarenogin is a natural compound extracted from toad species with unknown details.
