CAS RN 16956-91-5
CAS RN 16956-91-5
IUPAC Name:
trimethyl[2,3,5,6-tetrafluoro-4-(trimethylsilyl)phenyl]silane
Synonyms:
trimethyl[2,3,5,6-tetrafluoro-4-(trimethylsilyl)phenyl]silane
Molecular Weight (g/mol):
294.44
Molecular Formula:
C12H18F4Si2
InChi Key:
RIGHSQASRZUCPK-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)C1=C(F)C(F)=C(C(F)=C1F)[Si](C)(C)C