accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 169243-86-1

OHO(S)FNHOO

CAS RN 169243-86-1

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-fluorophenyl)propanoate
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-fluorophenyl)propanoate
Molecular Weight (g/mol): 404.42
Molecular Formula: C24H19FNO4
InChi Key: IXUMACXMEZBPJG-QFIPXVFZSA-M
SMILES: [O-]C(=O)[C@H](CC1=CC=C(F)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.