accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1655-07-8

H3COOO

CAS RN 1655-07-8

IUPAC Name: ethyl 2-oxocyclohexane-1-carboxylate
Synonyms: ethyl 2-oxocyclohexane-1-carboxylate
Molecular Weight (g/mol): 170.21
Molecular Formula: C9H14O3
InChi Key: FGSGHBPKHFDJOP-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCCCC1=O
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.