CAS RN 1572414-83-5
CAS RN 1572414-83-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Tocris ML 323
Potent and selective USP1-UAF1 allosteric inhibitor
2
Medchem Express ML-323, MedChemExpress
MedChemExpress ML-323 is a reversible, potent USP1-UAF1 inhibitor with IC50 of 76 nM in a Ub-Rho assay. The measured inhibition constants of ML-323 for the free enzyme (Ki) is 68 nM.

3
4