accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 15622-65-8

ClH3CH3CHNNHOO

CAS RN 15622-65-8

IUPAC Name: 4-[(3-ethyl-2-methyl-4-oxo-4,5,6,7-tetrahydro-1H-indol-5-yl)methyl]morpholin-4-ium chloride
Synonyms: 4-[(3-ethyl-2-methyl-4-oxo-1,5,6,7-tetrahydroindol-5-yl)methyl]morpholin-4-ium chloride
Molecular Weight (g/mol): 312.84
Molecular Formula: C16H25ClN2O2
InChi Key: GQWNECFJGBQMBO-UHFFFAOYNA-N
SMILES: [Cl-].CCC1=C(C)NC2=C1C(=O)C(C[NH+]1CCOCC1)CC2
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.