accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1552-42-7

CH3NH3COONCH3CH3NH3CCH3

CAS RN 1552-42-7

IUPAC Name: 6-(dimethylamino)-3,3-bis[4-(dimethylamino)phenyl]-1,3-dihydro-2-benzofuran-1-one
Synonyms: 6-(dimethylamino)-3,3-bis[4-(dimethylamino)phenyl]-2-benzofuran-1-one
Molecular Weight (g/mol): 415.54
Molecular Formula: C26H29N3O2
InChi Key: IPAJDLMMTVZVPP-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)C1(OC(=O)C2=CC(=CC=C12)N(C)C)C1=CC=C(C=C1)N(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.