accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 152918-18-8

H3CNHO(S)O(R)(R)OH(S)OHNNNHINN

CAS RN 152918-18-8

IUPAC Name: (2S,3S,4R,5R)-3,4-dihydroxy-5-(6-{[(3-iodophenyl)methyl]amino}-9H-purin-9-yl)-N-methyloxolane-2-carboxamide
Synonyms: (2S,3S,4R,5R)-3,4-dihydroxy-5-(6-{[(3-iodophenyl)methyl]amino}purin-9-yl)-N-methyloxolane-2-carboxamide
Molecular Weight (g/mol): 510.29
Molecular Formula: C18H19IN6O4
InChi Key: HUJXGQILHAUCCV-MOROJQBDSA-N
SMILES: CNC(=O)[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C(NCC3=CC=CC(I)=C3)N=CN=C12
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.