accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 15028-39-4

H3COONH2HCl

CAS RN 15028-39-4

IUPAC Name: methyl 2-amino-2-phenylacetate hydrochloride
Synonyms: methyl 2-amino-2-phenylacetate hydrochloride
Molecular Weight (g/mol): 201.65
Molecular Formula: C9H12ClNO2
InChi Key: DTHMTBUWTGVEFG-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(N)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.