accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 14526-22-8

OOOOOOCHHCCHH3CCH3CH3FeCF3CF3CF3

CAS RN 14526-22-8

IUPAC Name: iron(3+) 1,1,1-trifluoro-4-oxopent-2-en-2-olate
Synonyms: iron(3+) 1,1,1-trifluoro-4-oxopent-2-en-2-olate
Molecular Weight (g/mol): 208.92
Molecular Formula: C5H4F3FeO2
InChi Key: UDAOXISMUTYSIP-UHFFFAOYSA-M
SMILES: [Fe+3].CC(=O)C=C([O-])C(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.