accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 14024-18-1

FeOOOOOOH3CCH3CH3H3CH3CH3C

CAS RN 14024-18-1

IUPAC Name: iron(3+) tris(2,4-dioxopentan-3-ide)
Synonyms: tris(acetylacetonato)iron(III)
Molecular Weight (g/mol): 353.17
Molecular Formula: C15H21FeO6
InChi Key: KAMWVXQBCBPBND-UHFFFAOYSA-N
SMILES: [Fe+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.