accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 137330-13-3

CH3(R)OO(R)HO(S)CH3(S)O(R)O(R)CH3(S)OH(S)(R)HONH3CCH3(R)N(S)CH3(R)H3C(R)H3CO(R)O(R)O(R)CH3(R)OH(R)OCH3(R)OCH3CH3OHPOHOHO(E)(E)

CAS RN 137330-13-3

IUPAC Name: (4R,5S,6S,7R,9R,11E,13E,15R,16R)-6-{[(2R,3R,4S,5S,6R)-4-(dimethylamino)-3,5-dihydroxy-6-methyloxan-2-yl]oxy}-7-{2-[(3R,5S)-3,5-dimethylpiperidin-1-yl]ethyl}-16-ethyl-4-hydroxy-15-({[(2R,3R,4R,5R,6R)-5-hydroxy-3,4-dimethoxy-6-methyloxan-2-yl]oxy}methyl)-5,9,13-trimethyl-1-oxacyclohexadeca-11,13-diene-2,10-dione; phosphoric acid
Synonyms: phosphoric acid; tilmicosin
Molecular Weight (g/mol): 967.14
Molecular Formula: C46H83N2O17P
InChi Key: NESIVXZOSKKUDP-ARVJLQODSA-N
SMILES: OP(O)(O)=O.CC[C@H]1OC(=O)C[C@@H](O)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)[C@@H](O)[C@@H]([C@H]2O)N(C)C)[C@@H](CCN2C[C@@H](C)C[C@@H](C)C2)C[C@@H](C)C(=O)\C=C\C(\C)=C\[C@@H]1CO[C@@H]1O[C@H](C)[C@@H](O)[C@@H](OC)[C@H]1OC
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.