CAS RN 1259389-38-2
CAS RN 1259389-38-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express AMG 837 calcium hydrate, MedChemExpress
MedChemExpress AMG 837 calcium hydrate is a potent, orally bioavailable and partial agonist of GPR40/FFA1. AMG 837 calcium hydrate inhibits specific [3H]AMG 837 binding at the human FFA1 receptor with a pIC50 of 8.13. AMG 837 calcium hydrate could enhance insulin secretion and lower glucose levels in rodents.
