accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1185-55-3

CH3OSiCH3OH3COH3C

CAS RN 1185-55-3

IUPAC Name: trimethoxy(methyl)silane
Synonyms: methyltrimethoxysilane
Molecular Weight (g/mol): 136.22
Molecular Formula: C4H12O3Si
InChi Key: BFXIKLCIZHOAAZ-UHFFFAOYSA-N
SMILES: CO[Si](C)(OC)OC
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (6)

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.