accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 109-84-2

NH2NHHO

CAS RN 109-84-2

IUPAC Name: 2-hydrazinylethan-1-ol
Synonyms: 2-hydroxyethylhydrazine epa pesticide chemical code 055502 brombloom caswell no. 488A BOH HEH ethanolhydrazine omaflora
Molecular Weight (g/mol): 76.1
Molecular Formula: C2H8N2O
InChi Key: GBHCABUWWQUMAJ-UHFFFAOYSA-N
SMILES: NNCCO
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.