man

Test

CAS RN 108-64-5

H3COOCH3CH3

CAS RN 108-64-5

IUPAC Name: ethyl 3-methylbutanoate
Synonyms: ethyl isovalerate ethyl isovalerianate ethyl isovalerate (natural)
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: PPXUHEORWJQRHJ-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(C)C
Boiling Point 
  • (6)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results