man

Test

CAS RN 108-55-4

OOO

CAS RN 108-55-4

IUPAC Name: oxane-2,6-dione
Synonyms: glutaric anhydride
Molecular Weight (g/mol): 114.1
Molecular Formula: C5H6O3
InChi Key: VANNPISTIUFMLH-UHFFFAOYSA-N
SMILES: O=C1CCCC(=O)O1
Boiling Point 
  • (4)
  • (4)
Percent Purity 
  • (4)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (4)
  • (4)
  • (4)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)