man

Test

CAS RN 108-44-1

CH3H2N

CAS RN 108-44-1

IUPAC Name: 3-methylaniline
Synonyms: M-toluidine
Molecular Weight (g/mol): 107.16
Molecular Formula: C7H9N
InChi Key: JJYPMNFTHPTTDI-UHFFFAOYSA-N
SMILES: CC1=CC=CC(N)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)