man

Test

CAS RN 108-37-2

ClBr

CAS RN 108-37-2

IUPAC Name: 1-bromo-3-chlorobenzene
Synonyms: M-bromochlorobenzene
Molecular Weight (g/mol): 191.45
Molecular Formula: C6H4BrCl
InChi Key: JRGGUPZKKTVKOV-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Br)=C1
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)