man

Test

CAS RN 108-33-8

CH3SNH2NN

CAS RN 108-33-8

IUPAC Name: 5-methyl-1,3,4-thiadiazol-2-amine
Synonyms: 5-methyl-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol): 115.15
Molecular Formula: C3H5N3S
InChi Key: HMPUHXCGUHDVBI-UHFFFAOYSA-N
SMILES: CC1=NN=C(N)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results