man

Test

CAS RN 1072-83-9

H3CONH

CAS RN 1072-83-9

IUPAC Name: 1-(1H-pyrrol-2-yl)ethan-1-one
Synonyms: 2-acetylpyrrole 2-pyrrolylethanone pyrrole-α-methyl ketone
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: IGJQUJNPMOYEJY-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CN1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)