CAS RN 107149-56-4
CAS RN 107149-56-4
IUPAC Name:
chlorodiethyl(propan-2-yl)silane
Synonyms:
chlorodiethylisopropylsilane
Molecular Weight (g/mol):
164.75
Molecular Formula:
C7H17ClSi
InChi Key:
OSBPGFIPLLCQMO-UHFFFAOYSA-N
SMILES:
CC[Si](Cl)(CC)C(C)C