CAS RN 107073-27-8
CAS RN 107073-27-8
IUPAC Name:
[3-(1H-pyrrol-1-yl)thiophen-2-yl]methanol
Synonyms:
[3-(pyrrol-1-yl)thiophen-2-yl]methanol
Molecular Weight (g/mol):
179.24
Molecular Formula:
C9H9NOS
InChi Key:
SMSUHWYHSLZKHS-UHFFFAOYSA-N
SMILES:
OCC1=C(C=CS1)N1C=CC=C1
1
–
1
of
1
results
1
3-(1-Pyrrolyl)-2-thiophenemethanol, 97%, Thermo Scientific™
CAS: 107073-27-8 Molecular Formula: C9H9NOS Molecular Weight (g/mol): 179.237 MDL Number: MFCD00052578 InChI Key: SMSUHWYHSLZKHS-UHFFFAOYSA-N Synonym: 3-pyrrol-1-yl thiophene-2-methanol,3-1h-pyrrol-1-yl-2-thienyl methanol,3-pyrrol-1-ylthiophen-2-yl methanol,3-1h-pyrrol-1-yl thiophen-2-yl methanol,3-pyrrol-1-yl thiophen-2-yl methanol,3-1h-pyrrol-1-yl thien-2-yl methanol,3-pyrrolyl-2-thienyl methan-1-ol,3-1-pyrrolyl-2-thiophenemethanol,3-1-pyrrolyl-2-thiophenyl methanol PubChem CID: 342464 IUPAC Name: (3-pyrrol-1-ylthiophen-2-yl)methanol SMILES: C1=CN(C=C1)C2=C(SC=C2)CO