man

Test

CAS RN 1070-89-9

NaH3CSiCH3H3CNSiCH3CH3H3C

CAS RN 1070-89-9

IUPAC Name: sodium bis(trimethylsilyl)azanide
Synonyms: sodium bis(trimethylsilyl)amide
Molecular Weight (g/mol): 183.38
Molecular Formula: C6H18NNaSi2
InChi Key: WRIKHQLVHPKCJU-UHFFFAOYSA-N
SMILES: [Na+].C[Si](C)(C)[N-][Si](C)(C)C
Molecular Weight (g/mol) 
  • (9)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (6)
  • (4)
  • (4)
  • (9)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (5)