man

Test

CAS RN 107-88-0

CH3OHHO

CAS RN 107-88-0

IUPAC Name: butane-1,3-diol
Synonyms: 1,3-butanediol 1,3 butylene glycol (R)-1,3-butylene glycol (r)-1,3-butylene glycol (S)-isomer 1,3-butylene glycol (R)-isomer 1,3-butylene glycol β-butylene glycol (s)-isomer 1,3-butylene glycol 1,3-butylene glycol (r)-isomer 1,3-butylene glycol
Molecular Weight (g/mol): 90.12
Molecular Formula: C4H10O2
InChi Key: PUPZLCDOIYMWBV-UHFFFAOYNA-N
SMILES: CC(O)CCO
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (5)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (4)
  • (4)
  • (1)