CAS RN 107-83-5
CAS RN 107-83-5
IUPAC Name:
2-methylpentane
Synonyms:
2-methylpentane
Molecular Weight (g/mol):
86.18
Molecular Formula:
C6H14
InChi Key:
AFABGHUZZDYHJO-UHFFFAOYSA-N
SMILES:
CCCC(C)C
1
–
4
of
4
results
1
2-Methylpentane, 99+%, Thermo Scientific Chemicals
CAS: 107-83-5 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00009406 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 IUPAC Name: 2-methylpentane SMILES: CCCC(C)C
2
2-Methylpentane, 99+%, pure, Thermo Scientific Chemicals
CAS: 107-83-5 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD00009406 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 IUPAC Name: 2-methylpentane SMILES: CCCC(C)C