man

Test

CAS RN 1069120-19-9

H3CH3CCH3OONHOH

CAS RN 1069120-19-9

IUPAC Name: tert-butyl N-[4-(2-hydroxyethyl)cyclohexyl]carbamate
Synonyms: tert-butyl N-[4-(2-hydroxyethyl)cyclohexyl]carbamate
Molecular Weight (g/mol): 243.35
Molecular Formula: C13H25NO3
InChi Key: KTNFSGIXLVLQNK-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CCC(CCO)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)