man

Test

CAS RN 1067-74-9

CH3OPOOOH3COH3C

CAS RN 1067-74-9

IUPAC Name: methyl 2-(diethoxyphosphoryl)acetate
Synonyms: methyl 2-(diethoxyphosphoryl)acetate
Molecular Weight (g/mol): 210.17
Molecular Formula: C7H15O5P
InChi Key: CTSAXXHOGZNKJR-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC(=O)OC)OCC
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)