man

Test

CAS RN 1067-42-1

H3CGeCH3H3CH3C

CAS RN 1067-42-1

IUPAC Name: tetrabutylgermane
Synonyms: tetrabutylgermane
Molecular Weight (g/mol): 301.09
Molecular Formula: C16H36Ge
InChi Key: HDVLQIDIYKIVRE-UHFFFAOYSA-N
SMILES: CCCC[Ge](CCCC)(CCCC)CCCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results