man

Test

CAS RN 106463-17-6

ClCH3OOHNCH3OCH3SNH2OOH

CAS RN 106463-17-6

IUPAC Name: hydrogen 5-[(2R)-2-{[2-(2-ethoxyphenoxy)ethyl]amino}propyl]-2-methoxybenzene-1-sulfonamide chloride
Synonyms: flomax tamsulosin hydrochloride harnal pradif omnic
Molecular Weight (g/mol): 444.97
Molecular Formula: C20H29ClN2O5S
InChi Key: ZZIZZTHXZRDOFM-XFULWGLBSA-N
SMILES: [H+].[Cl-].CCOC1=CC=CC=C1OCCN[C@H](C)CC1=CC=C(OC)C(=C1)S(N)(=O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results