CAS RN 106391-87-1
CAS RN 106391-87-1
IUPAC Name:
tert-butyl N-[(2R)-1-hydroxy-3-methylbutan-2-yl]carbamate
Synonyms:
tert-butyl N-[(2R)-1-hydroxy-3-methylbutan-2-yl]carbamate
Molecular Weight (g/mol):
203.28
Molecular Formula:
C10H21NO3
InChi Key:
OOQRRYDVICNJGC-QMMMGPOBSA-N
SMILES:
CC(C)[C@H](CO)NC(=O)OC(C)(C)C
1
–
2
of
2
results
2
N-Boc-D-valinol, 95%, Thermo Scientific™
CAS: 106391-87-1 Molecular Formula: C10H21NO3 Molecular Weight (g/mol): 203.28 MDL Number: MFCD00235960 InChI Key: OOQRRYDVICNJGC-QMMMGPOBSA-N Synonym: boc-d-valinol,n-boc-d-valinol,r-tert-butyl 1-hydroxy-3-methylbutan-2-yl carbamate,tert-butyl n-2r-1-hydroxy-3-methylbutan-2-yl carbamate,boc-d-val-ol,tert-butyl n-1r-1-hydroxymethyl-2-methyl-propyl carbamate,n-tert-butoxycarbonyl-d-valinol,n-t-boc-d-valinol,ksc491q4r PubChem CID: 11241137 IUPAC Name: tert-butyl N-[(2R)-1-hydroxy-3-methylbutan-2-yl]carbamate SMILES: CC(C)[C@H](CO)NC(=O)OC(C)(C)C