man

Test

CAS RN 106266-06-2

H3CNNOFONN

CAS RN 106266-06-2

IUPAC Name: 3-{2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl}-2-methyl-4H,6H,7H,8H,9H-pyrido[1,2-a]pyrimidin-4-one
Synonyms: risperidone risperidal nu-buspirone risperdal M-tab bespar gen-buspirone apo-buspirone risperidone (product) buspirone preparation risperidone (substance)
Molecular Weight (g/mol): 410.49
Molecular Formula: C23H27FN4O2
InChi Key: RAPZEAPATHNIPO-UHFFFAOYSA-N
SMILES: CC1=C(CCN2CCC(CC2)C2=NOC3=CC(F)=CC=C23)C(=O)N2CCCCC2=N1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)