man

Test

CAS RN 106-69-4

HOOHOH

CAS RN 106-69-4

IUPAC Name: hexane-1,2,6-triol
Synonyms: 1,2,6-hexanetriol
Molecular Weight (g/mol): 134.18
Molecular Formula: C6H14O3
InChi Key: ZWVMLYRJXORSEP-UHFFFAOYNA-N
SMILES: OCCCCC(O)CO
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)