man

Test

CAS RN 106-52-5

CH3NOH

CAS RN 106-52-5

IUPAC Name: 1-methylpiperidin-4-ol
Synonyms: 4-piperidinol, 1-methyl-
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: BAUWRHPMUVYFOD-UHFFFAOYSA-N
SMILES: CN1CCC(O)CC1
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)