man

Test

CAS RN 106-22-9

CH3HOCH3CH3

CAS RN 106-22-9

IUPAC Name: (3R)-3,7-dimethyloct-6-en-1-ol
Synonyms: β-citronellol (R)-(+)-citronellol
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: QMVPMAAFGQKVCJ-SNVBAGLBSA-N
SMILES: C[C@@H](CCO)CCC=C(C)C
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)