CAS RN 1053058-99-3
CAS RN 1053058-99-3
IUPAC Name:
N'-hydroxy-3,4-dimethoxybenzene-1-carboximidamide
Synonyms:
N'-hydroxy-3,4-dimethoxybenzenecarboximidamide
Molecular Weight (g/mol):
196.21
Molecular Formula:
C9H12N2O3
InChi Key:
REKFOHFJHBBXOR-UHFFFAOYSA-N
SMILES:
COC1=C(OC)C=C(C=C1)C(N)=NO
1
–
1
of
1
results
1
3,4-Dimethoxybenzamidoxime, 97%, Thermo Scientific™
CAS: 1053058-99-3 Molecular Formula: C9H12N2O3 Molecular Weight (g/mol): 196.21 MDL Number: MFCD06147747,MFCD09842202 InChI Key: REKFOHFJHBBXOR-UHFFFAOYSA-N Synonym: 3,4-dimethoxybenzamidoxime,z-n'-hydroxy-3,4-dimethoxybenzene-1-carboximidamide,3,4-dimethoxyphenyl hydroxyimino methylamine,n'-hydroxy-3,4-dimethoxybenzimidamide,benzenecarboximidamide, n-hydroxy-3,4-dimethoxy,n'-hydroxy-3,4-dimethoxybenzene-1-carboximidamide,n'∼1∼-hydroxy-3,4-dimethoxy-1-benzenecarboximidamide,e-n'-hydroxy-3,4-dimethoxybenzenecarboximidamide PubChem CID: 9723450 IUPAC Name: N'-hydroxy-3,4-dimethoxybenzenecarboximidamide SMILES: COC1=C(OC)C=C(C=C1)C(N)=NO