man

Test

CAS RN 105-76-0

H3COOOOCH3

CAS RN 105-76-0

IUPAC Name: 1,4-dibutyl (2Z)-but-2-enedioate
Synonyms: dibutyl maleate
Molecular Weight (g/mol): 228.29
Molecular Formula: C12H20O4
InChi Key: JBSLOWBPDRZSMB-FPLPWBNLSA-N
SMILES: CCCCOC(=O)\C=C/C(=O)OCCCC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)